missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoro-4-methylphenylacetic acid, 97%

$44.58 - $109.95

Chemical Identifiers

CAS 261763-47-7
Molecular Formula C9H8F2O2
Formula Weight 186.16
MDL Number MFCD01631649
Chemical Name or Material 2,3-Difluoro-4-methylphenylacetic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2623503 SDS Alfa Aesar™
H2623503
1g Each for $109.95
AAH26235MD SDS Alfa Aesar™
H26235MD
250mg Each for $44.58
Specifications Description & Specifications

Chemical Identifiers

261763-47-7
186.16
2,3-Difluoro-4-methylphenylacetic acid
C9H8F2O2
MFCD01631649

Specifications

124° to 126°C
261763-47-7
186.16
2,3-Difluoro-4-methylphenylacetic acid
1g
C9H8F2O2
MFCD01631649
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning