Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoro-4-methylbenzamide, 97+%

$47.46 - $156.70

Chemical Identifiers

Chemical Name or Material 2,3-Difluoro-4-methylbenzamide
CAS 261763-35-3
Molecular Formula C8H7F2NO
Formula Weight 171.15
MDL Number MFCD01631633
Synonym 2,3-Difluoro-p-toluamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2425403

  SDS
alfa aesar™
B2425403
1g Each for $47.46

AAB2425406

  SDS
alfa aesar™
B2425406
5g Each for $156.70
Description & Specifications

Specifications

Formula Weight 171.15
MDL Number MFCD01631633
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥97%
Synonym 2,3-Difluoro-p-toluamide
Health Hazard 1 Warning
CAS 261763-35-3
Chemical Name or Material 2,3-Difluoro-4-methylbenzamide
Quantity 1g
Melting Point 158° to 162°C
Molecular Formula C8H7F2NO
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.