Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoro-4-methylaniline, 97%

$187.80 - $561.84

Chemical Identifiers

Chemical Name or Material 2,3-Difluoro-4-methylaniline
CAS 886503-79-3
Molecular Formula C7H7F2N
Formula Weight 143.14
MDL Number MFCD06660321
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2673103

  SDS
alfa aesar™
H2673103
1g Each for $187.80

AAH2673106

  SDS
alfa aesar™
H2673106
5g Each for $561.84
Description & Specifications

Specifications

Formula Weight 143.14
Chemical Name or Material 2,3-Difluoro-4-methylaniline
Assay 97%
Molecular Formula C7H7F2N
MDL Number MFCD06660321
CAS 886503-79-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.