Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoro-4-methoxyphenol, 97%

$266.70 - $803.11

Chemical Identifiers

Chemical Name or Material 2,3-Difluoro-4-methoxyphenol
CAS 261763-29-5
Molecular Formula C7H6F2O2
Formula Weight 160.12
MDL Number MFCD01631505
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2630503

  SDS
alfa aesar™
H2630503
1g Each for $266.70

AAH2630506

  SDS
alfa aesar™
H2630506
5g Each for $803.11
Description & Specifications

Specifications

Formula Weight 160.12
MDL Number MFCD01631505
Health Hazard 2 GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Assay 97%
Chemical Name or Material 2,3-Difluoro-4-methoxyphenol
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
CAS 261763-29-5
Melting Point 103° to 106°C
Quantity 1g
Molecular Formula C7H6F2O2
Health Hazard 1 Danger

Applications
Analytical application of poly{methyl[3-(2-hydroxy-3,4-difluoro)phenyl]propyl siloxane} as a QCM coating for DMMP detection.

Solubility
Slightly Soluble (1.8 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.