missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoro-4-methoxybenzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H5325103

 View more versions of this product

Catalog No. AAH5325103



Specifications

Specifications

2,3-Difluoro-4-methoxybenzeneboronic acid
170981-41-6
MFCD07368234
HIWYFMBSFSTYGU-UHFFFAOYSA-N
(2,3-difluoro-4-methoxyphenyl)boronic acid
25067272
98%
1g
C7H7BF2O3
2,3-difluoro-4-methoxyphenyl boronic acid, 2,3-difluoro-4-methoxybenzeneboronic acid, 2,3-difluoro-4-methyloxyphenylboronic acid, boronic acid, 2,3-difluoro-4-methoxyphenyl, boronic acid, b-2,3-difluoro-4-methoxyphenyl, 2,3-difluoro-4-methoxyphenylboronicacid, 4-borono-2,3-difluoroanisole, 2,3-difluoroanisylboronic acid, 2,3-difluoro-4-methoxyphenylboronic aicd, 2,3-difluoro-4-methoxyphenyl boronicacid
B(C1=C(C(=C(C=C1)OC)F)F)(O)O
187.937
187.94

Chemical Identifiers

170981-41-6
187.937
HIWYFMBSFSTYGU-UHFFFAOYSA-N
25067272
B(C1=C(C(=C(C=C1)OC)F)F)(O)O
C7H7BF2O3
MFCD07368234
2,3-difluoro-4-methoxyphenyl boronic acid, 2,3-difluoro-4-methoxybenzeneboronic acid, 2,3-difluoro-4-methyloxyphenylboronic acid, boronic acid, 2,3-difluoro-4-methoxyphenyl, boronic acid, b-2,3-difluoro-4-methoxyphenyl, 2,3-difluoro-4-methoxyphenylboronicacid, 4-borono-2,3-difluoroanisole, 2,3-difluoroanisylboronic acid, 2,3-difluoro-4-methoxyphenylboronic aicd, 2,3-difluoro-4-methoxyphenyl boronicacid
(2,3-difluoro-4-methoxyphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents