Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoro-4-methoxybenzaldehyde, 97%

$103.07 - $578.59

Chemical Identifiers

Chemical Name or Material 2,3-Difluoro-4-methoxybenzaldehyde
CAS 256417-11-5
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD04115981
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2654503

  SDS
alfa aesar™
H2654503
1g Each for $103.07

AAH2654509

  SDS
alfa aesar™
H2654509
10g Each for $578.59
Description & Specifications

Specifications

Formula Weight 172.13
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2,3-Difluoro-4-methoxybenzaldehyde
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 256417-11-5
Quantity 1g
MDL Number MFCD04115981
Assay 97%
Molecular Formula C8H6F2O2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.