Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoro-4-methoxyaniline, 97%

Chemical Identifiers

Chemical Name or Material 2,3-Difluoro-4-methoxyaniline
CAS 155020-51-2
Molecular Formula C7H7F2NO
Formula Weight 159.14
MDL Number MFCD04115986
UN Number UN2811
Synonym 2,3-Difluoro-p-anisidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2628103

 
alfa aesar™
H2628103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2628106

 
alfa aesar™
H2628106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 159.14
Chemical Name or Material 2,3-Difluoro-4-methoxyaniline
Assay 97%
Molecular Formula C7H7F2NO
MDL Number MFCD04115986
CAS 155020-51-2
UN Number UN2811
Synonym 2,3-Difluoro-p-anisidine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 38° to 40°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.