Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoro-4-ethoxybenzeneboronic acid, 98%

Chemical Identifiers

Chemical Name or Material 2,3-Difluoro-4-ethoxybenzeneboronic acid
CAS 212386-71-5
Formula Weight 201.96
MDL Number MFCD09258743
Beilstein 19662857
Synonym 2,3-Difluoro-4-ethoxyphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6440803

  SDS
alfa aesar™
H6440803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6440806

  SDS
alfa aesar™
H6440806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 151° to 154°C
Formula Weight 201.96
MDL Number MFCD09258743
Beilstein 19662857
CAS 212386-71-5
Chemical Name or Material 2,3-Difluoro-4-ethoxybenzeneboronic acid
Assay 98%
Synonym 2,3-Difluoro-4-ethoxyphenylboronic acid
Quantity 1g
Physical Form Solid