Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Diethylpyrazine, 98%

$29.08 - $372.18

Chemical Identifiers

Chemical Name or Material 2,3-Diethylpyrazine
CAS 15707-24-1
Molecular Formula C8H12N2
Formula Weight 136.2
MDL Number MFCD00006151
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1327706

  SDS
alfa aesar™
A1327706
5g Each for $29.08

AAA1327722

  SDS
alfa aesar™
A1327722
100g Each for $372.18

AAA1327714

  SDS
alfa aesar™
A1327714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2,3-Diethylpyrazine
Flash Point 71°C (160°F)
Odor Stench
Boiling Point 180° to 182°C
Quantity 5g
CAS 15707-24-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 98%
Molecular Formula C8H12N2
Formula Weight 136.2
MDL Number MFCD00006151
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.