Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Diethyl-5-methylpyrazine, 98+%

$49.73 - $461.07

Chemical Identifiers

Chemical Name or Material 2,3-Diethyl-5-methylpyrazine
CAS 18138-04-0
Molecular Formula C9H14N2
Formula Weight 150.23
MDL Number MFCD00009635
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1568606

  SDS
alfa aesar™
A1568606
5g Each for $49.73

AAA1568614

  SDS
alfa aesar™
A1568614
25g Each for $132.45

AAA1568622

  SDS
alfa aesar™
A1568622
100g Each for $461.07
Description & Specifications

Specifications

Formula Weight 150.23
MDL Number MFCD00009635
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay ≥98%
Boiling Point 190°C
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Chemical Name or Material 2,3-Diethyl-5-methylpyrazine
Molecular Formula C9H14N2
Flash Point 80°C (176°F)
CAS 18138-04-0
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.