missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Diethyl-5-methylpyrazine, 98+%

Manufacturer:  Alfa Aesar™ A1568614

 View more versions of this product

Catalog No. AAA1568614



Specifications

Specifications

2,3-Diethyl-5-methylpyrazine
190°C
18138-04-0
MFCD00009635
PSINWXIDJYEXLO-UHFFFAOYSA-N
2,3-diethyl-5-methylpyrazine
28905
≥98%
80°C (176°F)
25g
C9H14N2
pyrazine, 2,3-diethyl-5-methyl, 2-methyl-5,6-diethylpyrazine, 2,3-diethyl-6-methylpyrazine, 5-methyl-2,3-diethylpyrazine, 2,3-diethyl-5-methyl-pyrazine, fema no. 3336, hazelnut pyrazine, pubchem8621, acmc-1ca9d, ksc491g2h
CCC1=NC=C(N=C1CC)C
150.225
150.23

Chemical Identifiers

18138-04-0
150.225
PSINWXIDJYEXLO-UHFFFAOYSA-N
28905
CCC1=NC=C(N=C1CC)C
C9H14N2
MFCD00009635
pyrazine, 2,3-diethyl-5-methyl, 2-methyl-5,6-diethylpyrazine, 2,3-diethyl-6-methylpyrazine, 5-methyl-2,3-diethylpyrazine, 2,3-diethyl-5-methyl-pyrazine, fema no. 3336, hazelnut pyrazine, pubchem8621, acmc-1ca9d, ksc491g2h
2,3-diethyl-5-methylpyrazine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents