missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',3'-Dideoxyguanosine, 90+%

$136.34 - $351.61

Chemical Identifiers

CAS 85326-06-3
Molecular Formula C10H13N5O3
Molecular Weight (g/mol) 251.246
MDL Number MFCD00057074
InChI Key OCLZPNCLRLDXJC-NTSWFWBYSA-N
Synonym 2',3'-dideoxyguanosine, guanosine, 2',3'-dideoxy, 2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, dideoxyguanosine, 2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6-one, deoxy-deoxy-guanosine, 2,3-didioxyguanosine, 2', 3'-dideoxyguanosine, 2,3-dideoxyguanosine, chembl78101
PubChem CID 64974
IUPAC Name 2-amino-9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES C1CC(OC1CO)N2C=NC3=C2NC(=NC3=O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65810MB View Documents Alfa Aesar™
J65810MB
25mg Each for $136.34
 
AAJ65810MF View Documents Alfa Aesar™
J65810MF
50mg Each for $217.43
 
AAJ65810MC View Documents Alfa Aesar™
J65810MC
100mg Each for $351.61
 
Specifications

Chemical Identifiers

85326-06-3
251.246
OCLZPNCLRLDXJC-NTSWFWBYSA-N
64974
C1CC(OC1CO)N2C=NC3=C2NC(=NC3=O)N
C10H13N5O3
MFCD00057074
2',3'-dideoxyguanosine, guanosine, 2',3'-dideoxy, 2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, dideoxyguanosine, 2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6-one, deoxy-deoxy-guanosine, 2,3-didioxyguanosine, 2', 3'-dideoxyguanosine, 2,3-dideoxyguanosine, chembl78101
2-amino-9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one

Specifications

White
85326-06-3
MFCD00057074
OCLZPNCLRLDXJC-NTSWFWBYSA-N
2-amino-9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
64974
Powder
2',3'-Dideoxyguanosine
25mg
C10H13N5O3
2',3'-dideoxyguanosine, guanosine, 2',3'-dideoxy, 2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, dideoxyguanosine, 2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6-one, deoxy-deoxy-guanosine, 2,3-didioxyguanosine, 2', 3'-dideoxyguanosine, 2,3-dideoxyguanosine, chembl78101
C1CC(OC1CO)N2C=NC3=C2NC(=NC3=O)N
251.246
251.24
≥90%
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C