missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2',3'-Dideoxyadenosine, 98%, Thermo Scientific Chemicals
$197.62 - $197.62
Chemical Identifiers
CAS | 4097-22-7 |
---|---|
Molecular Formula | C10H13N5O2 |
Molecular Weight (g/mol) | 235.25 |
MDL Number | MFCD00010534 |
InChI Key | WVXRAFOPTSTNLL-NKWVEPMBSA-N |
Synonym | 2',3'-dideoxyadenosine, dideoxyadenosine, adenosine, 2',3'-dideoxy, ddado, 2s,5r-5-6-amino-9h-purin-9-yl tetrahydrofuran-2-yl methanol, ccris 428, unii-4q86ah641a, d2a, dsstox_cid_3771, dsstox_rid_77189 |
PubChem CID | 20039 |
IUPAC Name | [(2S,5R)-5-(6-aminopurin-9-yl)oxolan-2-yl]methanol |
SMILES | NC1=C2N=CN([C@H]3CC[C@@H](CO)O3)C2=NC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ64573MB
|
Thermo Scientific Chemicals
J64573MB |
25 mg |
Each for $197.62
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4097-22-7 | |
235.25 | |
WVXRAFOPTSTNLL-NKWVEPMBSA-N | |
20039 | |
NC1=C2N=CN([C@H]3CC[C@@H](CO)O3)C2=NC=N1 |
C10H13N5O2 | |
MFCD00010534 | |
2',3'-dideoxyadenosine, dideoxyadenosine, adenosine, 2',3'-dideoxy, ddado, 2s,5r-5-6-amino-9h-purin-9-yl tetrahydrofuran-2-yl methanol, ccris 428, unii-4q86ah641a, d2a, dsstox_cid_3771, dsstox_rid_77189 | |
[(2S,5R)-5-(6-aminopurin-9-yl)oxolan-2-yl]methanol |
Specifications
4097-22-7 | |
White | |
MFCD00010534 | |
619924 | |
2',3'-dideoxyadenosine, dideoxyadenosine, adenosine, 2',3'-dideoxy, ddado, 2s,5r-5-6-amino-9h-purin-9-yl tetrahydrofuran-2-yl methanol, ccris 428, unii-4q86ah641a, d2a, dsstox_cid_3771, dsstox_rid_77189 | |
NC1=C2N=CN([C@H]3CC[C@@H](CO)O3)C2=NC=N1 | |
235.25 | |
235.24 | |
Powder |
181°C to 184°C | |
C10H13N5O2 | |
25 mg | |
14,3101 | |
WVXRAFOPTSTNLL-NKWVEPMBSA-N | |
[(2S,5R)-5-(6-aminopurin-9-yl)oxolan-2-yl]methanol | |
20039 | |
98% | |
2',3'-Dideoxyadenosine |
Safety and Handling
EINECSNumber : 223-853-2
RTECSNumber : AU7358900
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only