missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dicyanohydroquinone, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 232050050
Specifications
2, 3-Dicyanohydroquinone | |
Glass bottle | |
>230.0°C | |
98% | |
97.5 | |
97.5% min. (HPLC) | |
(NC)2C6H2-1,4-(OH)2 | |
1,2-benzenedicarbonitrile, 3,6-dihydroxy, 2,3-dicyano-p-hydroquinone, 2,3-dicyanobenzene-1,4-diol, 2,3-dicyanohydroquinme, 2,3-dicyanohydroquinone, 3,6-dihydroxyphthalodinitrile, 3,6-dihydroxyphthalonitrile, acmc-1asrw, dicyanohydroquinone, phthalonitrile, 3,6-dihydroxy | |
MPAIWVOBMLSHQA-UHFFFAOYSA-N | |
3,6-dihydroxybenzene-1,2-dicarbonitrile | |
78467 | |
Crystalline Powder or Needles |
Authentic | |
Brown to Khaki | |
5g | |
4733-50-0 | |
100.0 | |
C8H4N2O2 | |
MFCD00001790 | |
Solubility in water: soluble in hot water. Other solubilities: sparingly soluble in cold water,soluble in ether,acetone,ethanol,sparingly soluble in cold water,benzene,chlorofo,rm | |
C1=CC(=C(C(=C1O)C#N)C#N)O | |
160.13 | |
160.13 | |
98% |
Chemical Identifiers
4733-50-0 | |
160.13 | |
MPAIWVOBMLSHQA-UHFFFAOYSA-N | |
78467 | |
C1=CC(=C(C(=C1O)C#N)C#N)O |
C8H4N2O2 | |
MFCD00001790 | |
1,2-benzenedicarbonitrile, 3,6-dihydroxy, 2,3-dicyano-p-hydroquinone, 2,3-dicyanobenzene-1,4-diol, 2,3-dicyanohydroquinme, 2,3-dicyanohydroquinone, 3,6-dihydroxyphthalodinitrile, 3,6-dihydroxyphthalonitrile, acmc-1asrw, dicyanohydroquinone, phthalonitrile, 3,6-dihydroxy | |
3,6-dihydroxybenzene-1,2-dicarbonitrile |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic in contact with skin.
Causes skin irritation.
Toxic if inhaled.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fre
GHS Signal Word: Danger
EINECSNumber : 225-241-0
TSCA : TSCA