missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichlorotoluene 98.0+%, TCI America™
$98.79 - $708.33
Chemical Identifiers
| CAS | 32768-54-0 |
|---|---|
| Molecular Formula | C7H6Cl2 |
| Molecular Weight (g/mol) | 161.025 |
| MDL Number | MFCD00000546 |
| InChI Key | GWLKCPXYBLCEKC-UHFFFAOYSA-N |
| Synonym | 2,3-dichlorotoluene, benzene, 1,2-dichloro-3-methyl, toluene, 2,3-dichloro, 1,2-dichloro-3-methyl-benzene, acmc-209hvi, ksc490m7b, gwlkcpxyblcekc-uhfffaoysa, 2,3-dct, #, labotest-bb ltbb004413 |
| PubChem CID | 34702 |
| IUPAC Name | 1,2-dichloro-3-methylbenzene |
| SMILES | CC1=C(C(=CC=C1)Cl)Cl |
Chemical Identifiers
| 32768-54-0 | |
| 161.025 | |
| GWLKCPXYBLCEKC-UHFFFAOYSA-N | |
| 34702 | |
| CC1=C(C(=CC=C1)Cl)Cl |
| C7H6Cl2 | |
| MFCD00000546 | |
| 2,3-dichlorotoluene, benzene, 1,2-dichloro-3-methyl, toluene, 2,3-dichloro, 1,2-dichloro-3-methyl-benzene, acmc-209hvi, ksc490m7b, gwlkcpxyblcekc-uhfffaoysa, 2,3-dct, #, labotest-bb ltbb004413 | |
| 1,2-dichloro-3-methylbenzene |
Specifications
| 32768-54-0 | |
| 208°C | |
| MFCD00000546 | |
| 2,3-dichlorotoluene, benzene, 1,2-dichloro-3-methyl, toluene, 2,3-dichloro, 1,2-dichloro-3-methyl-benzene, acmc-209hvi, ksc490m7b, gwlkcpxyblcekc-uhfffaoysa, 2,3-dct, #, labotest-bb ltbb004413 | |
| CC1=C(C(=CC=C1)Cl)Cl | |
| 161.025 | |
| 161.03 | |
| Liquid |
| Colorless | |
| C7H6Cl2 | |
| 25 g | |
| GWLKCPXYBLCEKC-UHFFFAOYSA-N | |
| 1,2-dichloro-3-methylbenzene | |
| 34702 | |
| ≥98.0% (GC) | |
| 2,3-Dichlorotoluene |
Safety and Handling
EINECSNumber : (3)-0078
TSCA : Yes