Please login to your online account to display your discounted pricing

2,3-Dichlorotoluene, 98%, ACROS Organics™


Quantity 50mL

Manufacturer: thermo fisher scientific  209190500

 View more versions of this product

Catalog No. AC209190500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 161.03
Quantity 50mL
Color Clear colorless to slightly yellow
TSCA TSCA
EINECS Number 251-203-8
Density 1.22g/mL
Beilstein 05, 295
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Infrared Spectrum Authentic
Refractive Index 1.5500 to 1.5520 (20°C, 598nm)
MDL Number MFCD00000546
Linear Formula CH3C6H3Cl2
Chemical Name or Material 2, 3-Dichlorotoluene
CAS 32768-54-0
Assay Percent Range 98%
Molecular Formula C7H6Cl2
Name Note 98%

Specifications

Chemical Name or Material 2, 3-Dichlorotoluene
Name Note 98%
CAS 32768-54-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H6Cl2
Linear Formula CH3C6H3Cl2
Formula Weight 161.03
MDL Number MFCD00000546
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents