missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichlorotoluene, 98%, ACROS Organics™

$186.72 - $186.72

Chemical Identifiers

CAS 32768-54-0
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H6Cl2
Linear Formula CH3C6H3Cl2
Formula Weight 161.03
MDL Number MFCD00000546
Chemical Name or Material 2, 3-Dichlorotoluene, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC209190500 SDS Thermo Fisher Scientific
209190500
50mL Each for $186.72
Specifications Description & Specifications

Chemical Identifiers

32768-54-0
100.0
C7H6Cl2
161.03
2, 3-Dichlorotoluene, 98%
97.5
98%
CH3C6H3Cl2
MFCD00000546
≥97.5%

Specifications

50mL
97.5
1.22g/mL
1.5500 to 1.5520 (20°C, 598nm)
98%
CH3C6H3Cl2
MFCD00000546
05, 295
≥97.5%
32768-54-0
100.0
Authentic
Liquid
C7H6Cl2
161.03
Clear colorless to slightly yellow
2, 3-Dichlorotoluene, 98%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 251-203-8

TSCA : TSCA