missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichlorotoluene, 98%

Manufacturer:  Alfa Aesar™ A1120622

 View more versions of this product

Catalog No. AAA1120622



Specifications

Specifications

2,3-Dichlorotoluene
207°C to 208°C
100g
C7H6Cl2
UN3082
2,3-dichlorotoluene, benzene, 1,2-dichloro-3-methyl, toluene, 2,3-dichloro, 1,2-dichloro-3-methyl-benzene, acmc-209hvi, ksc490m7b, gwlkcpxyblcekc-uhfffaoysa, 2,3-dct, #, labotest-bb ltbb004413
CC1=C(C(=CC=C1)Cl)Cl
161.025
161.03
83°C (181°F)
4°C
32768-54-0
MFCD00000546
2412384
GWLKCPXYBLCEKC-UHFFFAOYSA-N
1,2-dichloro-3-methylbenzene
34702
98%

Chemical Identifiers

32768-54-0
161.025
GWLKCPXYBLCEKC-UHFFFAOYSA-N
34702
CC1=C(C(=CC=C1)Cl)Cl
C7H6Cl2
MFCD00000546
2,3-dichlorotoluene, benzene, 1,2-dichloro-3-methyl, toluene, 2,3-dichloro, 1,2-dichloro-3-methyl-benzene, acmc-209hvi, ksc490m7b, gwlkcpxyblcekc-uhfffaoysa, 2,3-dct, #, labotest-bb ltbb004413
1,2-dichloro-3-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : TSCA

SDS Documents