Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichlorothiophenol, 97%

$65.88 - $761.95

Chemical Identifiers

Chemical Name or Material 2,3-Dichlorothiophenol
CAS 17231-95-7
Molecular Formula C6H4Cl2S
Formula Weight 179.07
MDL Number MFCD00041420
Beilstein 2043616
Synonym 2,3-Dichlorobenzenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1549106

  SDS
alfa aesar™
A1549106
5g Each for $65.88

AAA1549114

  SDS
alfa aesar™
A1549114
25g Each for $229.35

AAA1549122

  SDS
alfa aesar™
A1549122
100g Each for $761.95
Description & Specifications

Specifications

Formula Weight 179.07
Chemical Name or Material 2,3-Dichlorothiophenol
Assay 97%
Molecular Formula C6H4Cl2S
MDL Number MFCD00041420
CAS 17231-95-7
Beilstein 2043616
Synonym 2,3-Dichlorobenzenethiol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Odor Unpleasant
Boiling Point 95° to 98°C (0.5mmHg)
Melting Point 55° to 58°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.