missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichlorothiophenol, 97%

Manufacturer:  Alfa Aesar™ A1549114

 View more versions of this product

Catalog No. AAA1549114



Specifications

Specifications

2,3-Dichlorothiophenol
95°C to 98°C (0.5mmHg)
25g
C6H4Cl2S
2043616
QGRKONUHHGBHRB-UHFFFAOYSA-N
2,3-dichlorobenzenethiol
140202
97%
Unpleasant
55°C to 58°C
17231-95-7
MFCD00041420
2,3-dichlorothiophenol, 2,3-dichlorobenzene-1-thiol, dichlorothiophenol, benzenethiol, dichloro, 2,3-dichloro thiophenol, pubchem6796, 2,3-dichiorothiophenol, 2,3-dichloro-thiophenol, acmc-1cf9p, 2,3-dichlorobenzene thiol
C1=CC(=C(C(=C1)Cl)Cl)S
179.058
179.07

Chemical Identifiers

17231-95-7
179.058
QGRKONUHHGBHRB-UHFFFAOYSA-N
140202
C1=CC(=C(C(=C1)Cl)Cl)S
C6H4Cl2S
MFCD00041420
2,3-dichlorothiophenol, 2,3-dichlorobenzene-1-thiol, dichlorothiophenol, benzenethiol, dichloro, 2,3-dichloro thiophenol, pubchem6796, 2,3-dichiorothiophenol, 2,3-dichloro-thiophenol, acmc-1cf9p, 2,3-dichlorobenzene thiol
2,3-dichlorobenzenethiol
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents