missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichlorothiophene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC258790010

 View more versions of this product

Catalog No. AC258790010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 3-Dichlorothiophene
78°C
Glass Bottle
1.47
Colorless to Yellow
1g
96.0
96% min. (GC)
2,3-bis chloranyl thiophene, 2,3-dichloro thiophene, 2,3-dichloro-thiophene, acmc-20a4oq, ksc332o3r, pubchem15351, thiophene, 2,3-dichloro, thiophene, dichloro
LVNVLQIXMBTMPH-UHFFFAOYSA-N
2,3-dichlorothiophene
140207
Liquid
1.4700g/mL
Authentic
1.5687
172°C to 174°C
-37°C
17249-79-5
100.0
C4H2Cl2S
Solubility in water: <0.2g/L (20 c)
C1=CSC(=C1Cl)Cl
153.02
153.02
97%

Chemical Identifiers

17249-79-5
153.02
2,3-bis chloranyl thiophene, 2,3-dichloro thiophene, 2,3-dichloro-thiophene, acmc-20a4oq, ksc332o3r, pubchem15351, thiophene, 2,3-dichloro, thiophene, dichloro
2,3-dichlorothiophene
C4H2Cl2S
LVNVLQIXMBTMPH-UHFFFAOYSA-N
140207
C1=CSC(=C1Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents