missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichlorothiophene, 97%, ACROS Organics™

Chemical Identifiers

CAS 17249-79-5
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C4H2Cl2S
Formula Weight 153.02
Chemical Name or Material 2, 3-Dichlorothiophene
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC258790010 SDS Thermo Fisher Scientific
AC258790010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC258790050 SDS Thermo Fisher Scientific
AC258790050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

17249-79-5
100.0
C4H2Cl2S
2, 3-Dichlorothiophene
96.0
97%
153.02
≥96.0%

Specifications

153.02
1g
1.4700g/mL
96.0
C4H2Cl2S
Authentic
≥96.0%
Liquid
Clear colorless to pale yellow
100.0
17249-79-5
97%
2, 3-Dichlorothiophene

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger