missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichlorothiophene, 97%

Manufacturer:  Alfa Aesar™ L1057803

 View more versions of this product

Catalog No. AAL1057803



Specifications

Specifications

2,3-Dichlorothiophene
173°C to 174°C
17249-79-5
MFCD00043890
1760
LVNVLQIXMBTMPH-UHFFFAOYSA-N
2,3-dichlorothiophene
140207
97%
70°C (158°F)
1g
C4H2Cl2S
UN2810
thiophene, 2,3-dichloro, thiophene, dichloro, pubchem15351, acmc-20a4oq, 2,3-dichloro thiophene, 2,3-dichloro-thiophene, 2,3-bis chloranyl thiophene, ksc332o3r
C1=CSC(=C1Cl)Cl
153.02
153.03

Chemical Identifiers

17249-79-5
153.02
LVNVLQIXMBTMPH-UHFFFAOYSA-N
140207
C1=CSC(=C1Cl)Cl
C4H2Cl2S
MFCD00043890
thiophene, 2,3-dichloro, thiophene, dichloro, pubchem15351, acmc-20a4oq, 2,3-dichloro thiophene, 2,3-dichloro-thiophene, 2,3-bis chloranyl thiophene, ksc332o3r
2,3-dichlorothiophene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H227-H302-H312
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

Documents