Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichlorothiophene, 97%

Chemical Identifiers

Chemical Name or Material 2,3-Dichlorothiophene
CAS 17249-79-5
Molecular Formula C4H2Cl2S
Formula Weight 153.03
MDL Number MFCD00043890
UN Number UN2810
Beilstein 1760
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1057806

  SDS
alfa aesar™
L1057806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1057803

  SDS
alfa aesar™
L1057803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.03
Chemical Name or Material 2,3-Dichlorothiophene
Molecular Formula C4H2Cl2S
MDL Number MFCD00043890
CAS 17249-79-5
UN Number UN2810
Health Hazard 2 GHS H Statement
H331-H227-H302-H312
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Quantity 5g
Assay 97%
Beilstein 1760
Health Hazard 1 Danger
Flash Point 70°C (158°F)
Boiling Point 173° to 174°C
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H227-H302-H312
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.