missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichlorothiophene-5-sulfonyl chloride, 97%

Manufacturer:  Alfa Aesar™ L1178003

 View more versions of this product

Catalog No. AAL1178003



Specifications

Specifications

2,3-Dichlorothiophene-5-sulfonyl chloride
1g
C4HCl3O2S2
UN3261
2,3-dichlorothiophene-5-sulfonyl chloride, 2,3-dichlorothiophene-5-sulphonyl chloride, 4,5-dichloro-2-thiophenesulfonyl chloride, 4,5-dichloro-thiophene-2-sulfonyl chloride, 2-thiophenesulfonyl chloride, 4,5-dichloro, pubchem5523, acmc-20aoh3, 4,5-dichloro 2-thienyl chlorosulfone, 2,3-dichlorothiophene-5-sulfonylchloride, 2-thiophenesulfonylchloride, 4,5-dichloro
C1=C(SC(=C1Cl)Cl)S(=O)(=O)Cl
251.52
251.54
54°C to 57°C
126714-85-0
MFCD00052092
161983
IVTWLTRKVRJPNG-UHFFFAOYSA-N
4,5-dichlorothiophene-2-sulfonyl chloride
518622
97%

Chemical Identifiers

126714-85-0
251.52
IVTWLTRKVRJPNG-UHFFFAOYSA-N
518622
C1=C(SC(=C1Cl)Cl)S(=O)(=O)Cl
C4HCl3O2S2
MFCD00052092
2,3-dichlorothiophene-5-sulfonyl chloride, 2,3-dichlorothiophene-5-sulphonyl chloride, 4,5-dichloro-2-thiophenesulfonyl chloride, 4,5-dichloro-thiophene-2-sulfonyl chloride, 2-thiophenesulfonyl chloride, 4,5-dichloro, pubchem5523, acmc-20aoh3, 4,5-dichloro 2-thienyl chlorosulfone, 2,3-dichlorothiophene-5-sulfonylchloride, 2-thiophenesulfonylchloride, 4,5-dichloro
4,5-dichlorothiophene-2-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Documents