Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichlorothiobenzamide, 97%

$67.81 - $201.91

Chemical Identifiers

Chemical Name or Material 2,3-Dichlorothiobenzamide
CAS 84863-83-2
Molecular Formula C7H5Cl2NS
Formula Weight 206.09
MDL Number MFCD00221163
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2676103

  SDS
alfa aesar™
H2676103
1g Each for $67.81

AAH2676106

  SDS
alfa aesar™
H2676106
5g Each for $201.91
Description & Specifications

Specifications

Formula Weight 206.09
Melting Point 126° to 128°C
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 84863-83-2
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Chemical Name or Material 2,3-Dichlorothiobenzamide
Assay 97%
Health Hazard 1 Warning
Quantity 1g
MDL Number MFCD00221163
Molecular Formula C7H5Cl2NS
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.