missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dichloropyridine, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 113720500
Specifications
2, 3-Dichloropyridine | |
Glass bottle | |
67.0°C to 70.0°C | |
2402-77-9 | |
C5H3Cl2N | |
20,II,152 | |
Solubility in water: slightly soluble | |
C1=CC(=C(N=C1)Cl)Cl | |
147.99 | |
147.99 | |
99% |
Authentic | |
White to Beige | |
50g | |
98.5% min. (GC) | |
(C5H3N)Cl2 | |
2,3-dichloro pyridine, 2,3-dichloro-pyridine, 2.3-dichloro-pyridine, acmc-209g7x, ccris 1726, dichloropyridine, pubchem1198, pyridine, 2,3-dichloro, pyridine, dichloro, pyridine,3-dichloro | |
MAKFMOSBBNKPMS-UHFFFAOYSA-N | |
2,3-dichloropyridine | |
16988 | |
Crystalline Powder |
Chemical Identifiers
2402-77-9 | |
147.99 | |
2,3-dichloro pyridine, 2,3-dichloro-pyridine, 2.3-dichloro-pyridine, acmc-209g7x, ccris 1726, dichloropyridine, pubchem1198, pyridine, 2,3-dichloro, pyridine, dichloro, pyridine,3-dichloro | |
2,3-dichloropyridine |
C5H3Cl2N | |
MAKFMOSBBNKPMS-UHFFFAOYSA-N | |
16988 | |
C1=CC(=C(N=C1)Cl)Cl |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 219-281-8