missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichloropyridine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC113722500

 View more versions of this product

Catalog No. AC113722500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 3-Dichloropyridine
Glass Bottle
67.0°C to 70.0°C
2402-77-9
C5H3Cl2N
20,II,152
Solubility in water: slightly soluble
C1=CC(=C(N=C1)Cl)Cl
147.99
147.99
99%
Authentic
White to Beige
250g
98.5% min. (GC)
(C5H3N)Cl2
2,3-dichloro pyridine, 2,3-dichloro-pyridine, 2.3-dichloro-pyridine, acmc-209g7x, ccris 1726, dichloropyridine, pubchem1198, pyridine, 2,3-dichloro, pyridine, dichloro, pyridine,3-dichloro
MAKFMOSBBNKPMS-UHFFFAOYSA-N
2,3-dichloropyridine
16988
Crystalline Powder

Chemical Identifiers

2402-77-9
147.99
2,3-dichloro pyridine, 2,3-dichloro-pyridine, 2.3-dichloro-pyridine, acmc-209g7x, ccris 1726, dichloropyridine, pubchem1198, pyridine, 2,3-dichloro, pyridine, dichloro, pyridine,3-dichloro
2,3-dichloropyridine
C5H3Cl2N
MAKFMOSBBNKPMS-UHFFFAOYSA-N
16988
C1=CC(=C(N=C1)Cl)Cl
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 219-281-8

SDS Documents