missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dichloropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6373303
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,3-Dichloropyridine-4-carboxylic acid | |
203°C to 207°C | |
MFCD01861977 | |
2,3-dichloroisonicotinic acid, dichloroisonicotinic acid, 4-carboxy-2,3-dichloropyridine, 2,3-dichloro-4-pyridinecarboxylic acid, 2,3-dichloropyridine-4-carboxylicacid, 2,3-dichloro pyridine-4-carboxylic acid, pubchem22306, acmc-1cbyj, 2,3-dichloroisonicotinicacid, 2,3-dichloro-isonicotinic acid | |
OC(=O)C1=C(Cl)C(Cl)=NC=C1 | |
192.00 | |
192 | |
Solid |
184416-84-0 | |
C6H3Cl2NO2 | |
1 g | |
VGKZZKKTZKRCPA-UHFFFAOYSA-N | |
2,3-dichloropyridine-4-carboxylic acid | |
2736083 | |
97% |
Chemical Identifiers
184416-84-0 | |
192.00 | |
VGKZZKKTZKRCPA-UHFFFAOYSA-N | |
2736083 | |
OC(=O)C1=C(Cl)C(Cl)=NC=C1 |
C6H3Cl2NO2 | |
MFCD01861977 | |
2,3-dichloroisonicotinic acid, dichloroisonicotinic acid, 4-carboxy-2,3-dichloropyridine, 2,3-dichloro-4-pyridinecarboxylic acid, 2,3-dichloropyridine-4-carboxylicacid, 2,3-dichloro pyridine-4-carboxylic acid, pubchem22306, acmc-1cbyj, 2,3-dichloroisonicotinicacid, 2,3-dichloro-isonicotinic acid | |
2,3-dichloropyridine-4-carboxylic acid |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only