Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichloropyridine-4-boronic acid, 95%

$161.62 - $361.73

Chemical Identifiers

Chemical Name or Material 2,3-Dichloropyridine-4-boronic acid
CAS 951677-39-7
Molecular Formula C5H4BCl2NO2
Formula Weight 191.81
MDL Number MFCD04038756
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2766803

 
alfa aesar™
H2766803
1g Each for $361.73

AAH27668MD

 
alfa aesar™
H27668MD
250mg Each for $161.62
Description & Specifications

Specifications

Formula Weight 191.81
Chemical Name or Material 2,3-Dichloropyridine-4-boronic acid
Assay 95%
Molecular Formula C5H4BCl2NO2
MDL Number MFCD04038756
CAS 951677-39-7
Melting Point 136° to 138°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
It is used in HPLC and NMR chromatography. It is involved in several cross-coupling reactions.

Solubility
Soluble in organic solvents.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.