missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichloropyrazine, 98%

Manufacturer:  Alfa Aesar™ L1150006

 View more versions of this product

Catalog No. AAL1150006



Specifications

Specifications

2,3-Dichloropyrazine
5g
C4H2Cl2N2
112628
MLCNOCRGSBCAGH-UHFFFAOYSA-N
2,3-dichloropyrazine
78575
98%
105°C to 107°C (50mmHg)
4858-85-9
MFCD00040964
pyrazine, 2,3-dichloro, 2,3-dichloro-pyrazine, 2,3-dichloro pyrazine, 2,3-dichloro-1,4-diazine, 2,3-dichlorpyrazin, dichloropyrazine, dichloro pyrazine, zlchem 169, 2 3-dichloropyrazine, 2,3 dichloropyrazine
C1=CN=C(C(=N1)Cl)Cl
148.974
148.98

Chemical Identifiers

4858-85-9
148.974
MLCNOCRGSBCAGH-UHFFFAOYSA-N
78575
C1=CN=C(C(=N1)Cl)Cl
C4H2Cl2N2
MFCD00040964
pyrazine, 2,3-dichloro, 2,3-dichloro-pyrazine, 2,3-dichloro pyrazine, 2,3-dichloro-1,4-diazine, 2,3-dichlorpyrazin, dichloropyrazine, dichloro pyrazine, zlchem 169, 2 3-dichloropyrazine, 2,3 dichloropyrazine
2,3-dichloropyrazine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents