Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichloropropionitrile, 94%

Chemical Identifiers

Chemical Name or Material 2,3-Dichloropropionitrile
CAS 2601-89-0
Molecular Formula ClCH2CHClCN
Formula Weight 123.96
MDL Number MFCD00013809
UN Number UN3276
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1743809

  SDS
alfa aesar™
A1743809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1743818

  SDS
alfa aesar™
A1743818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 123.96
Chemical Name or Material 2,3-Dichloropropionitrile
Assay 94%
Molecular Formula ClCH2CHClCN
MDL Number MFCD00013809
CAS 2601-89-0
UN Number UN3276
Boiling Point 62° to 63°C (13mmHg)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300
Fatal if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300
Fatal if swallowed.