Learn More
2,3-Dichlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1071714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,3-Dichlorophenylhydrazine hydrochloride | |
∼204°C (decomposition) | |
MFCD00052265 | |
3708910 | |
HPIALSXRVQRGMK-UHFFFAOYSA-N | |
(2,3-dichlorophenyl)hydrazine;hydrochloride | |
12276572 | |
97% |
21938-47-6 | |
C6H6Cl2N2 | |
25 g | |
2,3-dichlorophenylhydrazine hydrochloride, 2,3-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,3-dichlorophenyl-, monohydrochloride, 1-2,3-dichlorophenyl hydrazine hydrochloride, pubchem7561, acmc-209fpn, ksc494k0t, 2,3-dichlorophenylhydrazine, chloride, 2,3-dichlorophenylhydrazine hcl | |
NNC1=CC=CC(Cl)=C1Cl | |
177.03 | |
213.49 |
Chemical Identifiers
21938-47-6 | |
177.03 | |
HPIALSXRVQRGMK-UHFFFAOYSA-N | |
12276572 | |
NNC1=CC=CC(Cl)=C1Cl |
C6H6Cl2N2 | |
MFCD00052265 | |
2,3-dichlorophenylhydrazine hydrochloride, 2,3-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,3-dichlorophenyl-, monohydrochloride, 1-2,3-dichlorophenyl hydrazine hydrochloride, pubchem7561, acmc-209fpn, ksc494k0t, 2,3-dichlorophenylhydrazine, chloride, 2,3-dichlorophenylhydrazine hcl | |
(2,3-dichlorophenyl)hydrazine;hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only