missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichlorophenyl Isocyanate, TCI America™

$36.67 - $111.53

Chemical Identifiers

CAS 41195-90-8
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.007
MDL Number MFCD00002000
InChI Key FYWJWWMKCARWQG-UHFFFAOYSA-N
Synonym 2,3-dichlorophenyl isocyanate, 2,3-dichlorophenylisocyanate, benzene, 1,2-dichloro-3-isocyanato, 1,2-dichloro-3-isocyanato-benzene, 2,3-dichlorobenzenisocyanate, benzene, dichloroisocyanato, dichlorphenylisocyanate, dichlorophenyl isocyanate, acmc-1ap4f, 2,3dichlorophenylisocyanate
PubChem CID 33050
IUPAC Name 1,2-dichloro-3-isocyanatobenzene
SMILES C1=CC(=C(C(=C1)Cl)Cl)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I04991G View Documents TCI America
I04991G
1g Each for $36.67
 
I04995G View Documents TCI America
I04995G
5g Each for $111.53
 
Specifications

Chemical Identifiers

41195-90-8
188.007
FYWJWWMKCARWQG-UHFFFAOYSA-N
33050
C1=CC(=C(C(=C1)Cl)Cl)N=C=O
C7H3Cl2NO
MFCD00002000
2,3-dichlorophenyl isocyanate, 2,3-dichlorophenylisocyanate, benzene, 1,2-dichloro-3-isocyanato, 1,2-dichloro-3-isocyanato-benzene, 2,3-dichlorobenzenisocyanate, benzene, dichloroisocyanato, dichlorphenylisocyanate, dichlorophenyl isocyanate, acmc-1ap4f, 2,3dichlorophenylisocyanate
1,2-dichloro-3-isocyanatobenzene

Specifications

66°C
1g
C7H3Cl2NO
2250
FYWJWWMKCARWQG-UHFFFAOYSA-N
1,2-dichloro-3-isocyanatobenzene
33050
Clear Liquid at 20°C
2,3-Dichlorophenyl Isocyanate
Yellow
41195-90-8
MFCD00002000
2,3-dichlorophenyl isocyanate, 2,3-dichlorophenylisocyanate, benzene, 1,2-dichloro-3-isocyanato, 1,2-dichloro-3-isocyanato-benzene, 2,3-dichlorobenzenisocyanate, benzene, dichloroisocyanato, dichlorphenylisocyanate, dichlorophenyl isocyanate, acmc-1ap4f, 2,3dichlorophenylisocyanate
C1=CC(=C(C(=C1)Cl)Cl)N=C=O
188.007
188.01
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator