Please login to your online account to display your discounted pricing

2,3-Dichlorophenyl isocyanate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3-Dichlorophenyl isocyanate
Name Note 97%
CAS 41195-90-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7 H3 Cl2 N O
Linear Formula Cl2C6H3NCO
Formula Weight 188.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC314250050

  SDS
thermo fisher scientific
314250050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid After Melting
Formula Weight 188.01
Quantity 5g
Color Clear colorless
TSCA TSCA
EINECS Number 255-253-1
Density 1.4240g/mL
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.
Toxic if swallowed.
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Refractive Index 1.5774 to 1.5794 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Linear Formula Cl2C6H3NCO
MDL Number MFCD00002000
Name Note 97%
Molecular Formula C7 H3 Cl2 N O
Chemical Name or Material 2, 3-Dichlorophenyl isocyanate
CAS 41195-90-8
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.
Toxic if swallowed.