Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichlorophenoxyacetic acid, 97%

$74.51 - $287.85

Chemical Identifiers

Chemical Name or Material 2,3-Dichlorophenoxyacetic acid
CAS 2976-74-1
Molecular Formula C8H6Cl2O3
Formula Weight 221.04
MDL Number MFCD00004299
Beilstein 1963475
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1323106

  SDS
alfa aesar™
A1323106
5g Each for $74.51

AAA1323114

  SDS
alfa aesar™
A1323114
25g Each for $287.85
Description & Specifications

Specifications

Formula Weight 221.04
Chemical Name or Material 2,3-Dichlorophenoxyacetic acid
Assay 97%
Molecular Formula C8H6Cl2O3
MDL Number MFCD00004299
CAS 2976-74-1
Beilstein 1963475
Melting Point 173° to 175°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.