Please login to your online account to display your discounted pricing

2,3-Dichlorophenethylamine, 99+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC371290050

 View more versions of this product

Catalog No. AC371290050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 190.07
Quantity 5g
Color Clear colorless to pale yellow
Density 1.2680g/mL
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 2, 3-Dichlorophenethylamine
Infrared Spectrum Authentic
Assay Percent Range 99+%
Molecular Formula C8H9Cl2N
Linear Formula Cl2C6H3CH2CH2NH2
MDL Number MFCD04112741
Packaging Glass bottle
CAS 34164-43-7
Refractive Index 1.4690 to 1.4710 (20°C, 589nm)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 3-Dichlorophenethylamine
CAS 34164-43-7
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H9Cl2N
Linear Formula Cl2C6H3CH2CH2NH2
Formula Weight 190.07
MDL Number MFCD04112741
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents