missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichlorophenethylamine, 99+%, ACROS Organics™

$111.01 - $111.01

Chemical Identifiers

CAS 34164-43-7
Molecular Formula C8H9Cl2N
Molecular Weight (g/mol) 190.07
MDL Number MFCD04112741
InChI Key WWRLSNWKOZHYIY-UHFFFAOYSA-N
Synonym 2,3-dichlorophenethylamine, 2-2,3,-dichlorophenyl ethylamine, 2-2,3-dichloro-phenyl-ethylamine, 2-2,3-dichlorophenyl ethan-1-amine, 2-2,3-dichlorophenyl ethanamine, 2-2,3-dichlorophenyl ethyl amine, 2-2,3-dichlorophenyl ethylamine, benzeneethanamine,2,3-dichloro
PubChem CID 2734758
IUPAC Name 2-(2,3-dichlorophenyl)ethanamine
SMILES C1=CC(=C(C(=C1)Cl)Cl)CCN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC371290010
SDS View Documents
Thermo Fisher Scientific
AC371290010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC371290050
SDS View Documents
Thermo Fisher Scientific
AC371290050
5g Glass bottle
Each for $111.01
 
AC371290250
SDS View Documents
Thermo Fisher Scientific
AC371290250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

34164-43-7
190.07
WWRLSNWKOZHYIY-UHFFFAOYSA-N
2734758
C1=CC(=C(C(=C1)Cl)Cl)CCN
C8H9Cl2N
MFCD04112741
2,3-dichlorophenethylamine, 2-2,3,-dichlorophenyl ethylamine, 2-2,3-dichloro-phenyl-ethylamine, 2-2,3-dichlorophenyl ethan-1-amine, 2-2,3-dichlorophenyl ethanamine, 2-2,3-dichlorophenyl ethyl amine, 2-2,3-dichlorophenyl ethylamine, benzeneethanamine,2,3-dichloro
2-(2,3-dichlorophenyl)ethanamine

Specifications

1.2680g/mL
Glass bottle
1.268
1g
99.0
99% min. (GC)
Cl2C6H3CH2CH2NH2
2,3-dichlorophenethylamine, 2-2,3,-dichlorophenyl ethylamine, 2-2,3-dichloro-phenyl-ethylamine, 2-2,3-dichlorophenyl ethan-1-amine, 2-2,3-dichlorophenyl ethanamine, 2-2,3-dichlorophenyl ethyl amine, 2-2,3-dichlorophenyl ethylamine, benzeneethanamine,2,3-dichloro
C1=CC(=C(C(=C1)Cl)Cl)CCN
190.07
190.07
99+%
Authentic
1.4690 to 1.4710 (20°C, 589nm)
Colorless to Yellow
34164-43-7
100.0
C8H9Cl2N
MFCD04112741
WWRLSNWKOZHYIY-UHFFFAOYSA-N
2-(2,3-dichlorophenyl)ethanamine
2734758
Liquid
2, 3-Dichlorophenethylamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Product Certifications