Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichlorooctafluorobutane, 97%

$34.35 - $100.96

Chemical Identifiers

Chemical Name or Material 2,3-Dichlorooctafluorobutane
CAS 355-20-4
Molecular Formula CF3CFClCFClCF3
Formula Weight 270.94
MDL Number MFCD00018064
Beilstein 1777520
Synonym CFC-318mbb
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0925203

  SDS
alfa aesar™
L0925203
1g Each for $34.35

AAL0925206

  SDS
alfa aesar™
L0925206
5g Each for $100.96
Description & Specifications

Specifications

Formula Weight 270.94
Molecular Formula CF3CFClCFClCF3
CAS 355-20-4
MDL Number MFCD00018064
Melting Point ≈−68°C
Beilstein 1777520
Assay 97%
Synonym CFC-318mbb
Boiling Point 62° to 63°C
Quantity 1g
Chemical Name or Material 2,3-Dichlorooctafluorobutane