missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichloronitrobenzene 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 113510010

 View more versions of this product

Catalog No. AC113510010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 3-Dichloronitrobenzene
Light yellow to light brown
1kg
3209-22-1
100.0
Crystalline Powder
99%
Cl2C6H3NO2
MFCD00007062
05, 245
1.449g/mL
60° to 63°C
99%
98.5
Authentic
≥98.5%
C6H3Cl2NO2
192
Glass bottle
1, 2-Dichloro-3-nitrobenzene

Chemical Identifiers

3209-22-1
100.0
C6H3Cl2NO2
192
1, 2-Dichloro-3-nitrobenzene
≥98.5%
98.5
99%
Cl2C6H3NO2
MFCD00007062
2, 3-Dichloronitrobenzene, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-717-7

RTECSNumber : CZ5240000

TSCA : TSCA