Please login to your online account to display your discounted pricing

2,3-Dichloronitrobenzene 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3-Dichloronitrobenzene
Name Note 99%
CAS 3209-22-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H3Cl2NO2
Linear Formula Cl2C6H3NO2
Formula Weight 192
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC113510010

  SDS
thermo fisher scientific
113510010
Glass bottle 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 192
Quantity 1kg
Color Light yellow to light brown
Beilstein 05, 245
Melting Point 60° to 63°C
EINECS Number 221-717-7
TSCA TSCA
RTECS Number CZ5240000
Density 1.449g/mL
CAS Max % 100.0
Packaging Glass bottle
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material 2, 3-Dichloronitrobenzene
Infrared Spectrum Authentic
Assay Percent Range 99%
CAS 3209-22-1
Synonym 1, 2-Dichloro-3-nitrobenzene
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99%
Molecular Formula C6H3Cl2NO2
Linear Formula Cl2C6H3NO2
MDL Number MFCD00007062