Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichlorobutane, (+/-) + meso, 99%

Chemical Identifiers

Chemical Name or Material 2,3-Dichlorobutane
CAS 7581-97-7
Name Note (+/-) + meso
Molecular Formula C4H8Cl2
Linear Formula CH3CHClCHClCH3
Formula Weight 127.01g/mol
MDL Number MFCD00000866
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2138606

 
alfa aesar™
B21386-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2138614

 
alfa aesar™
B21386-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Chloroform-like
Boiling Point 117° to 119°C
Melting Point −80°C
Flash Point 18°C (65°F)
Quantity 5g
Chemical Name or Material 2,3-Dichlorobutane
Name Note (+/-) + meso
CAS 7581-97-7
Assay 99%
Molecular Formula C4H8Cl2
Linear Formula CH3CHClCHClCH3
Formula Weight 127.01g/mol
MDL Number MFCD00000866
UN Number UN1993
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P261-P273-P243
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Take precautionary measures against static discharge.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P261-P273-P243
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Take precautionary measures against static discharge.

GHS H Statement
H225
Highly flammable liquid and vapour.