missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichlorobutane 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 113270250

 View more versions of this product

Catalog No. AC113270250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
127.01
Clear colorless to slightly yellow
01, 119
97.5
Authentic
98%
CH3CH(Cl)CH(Cl)CH3
98%
7581-97-7
Liquid
25mL
1.1000g/mL
100.0
1.4410 to 1.4430 (20°C, 589nm)
Glass bottle
mixture of DL and meso isomers
2, 3-Dichlorobutane
MFCD00000866
C4H8Cl2

Chemical Identifiers

7581-97-7
100.0
C4H8Cl2
127.01
2, 3-Dichlorobutane, 98%
97.5
98%
CH3CH(Cl)CH(Cl)CH3
MFCD00000866
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-486-4

TSCA : TSCA