missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichlorobutane 98%, ACROS Organics™

Chemical Identifiers

CAS 7581-97-7
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C4H8Cl2
Linear Formula CH3CH(Cl)CH(Cl)CH3
Formula Weight 127.01
MDL Number MFCD00000866
Chemical Name or Material 2, 3-Dichlorobutane, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC113270250 SDS Thermo Fisher Scientific
113270250
Glass bottle 25mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7581-97-7
100.0
C4H8Cl2
127.01
2, 3-Dichlorobutane, 98%
97.5
98%
CH3CH(Cl)CH(Cl)CH3
MFCD00000866
≥97.5%

Specifications

1.1000g/mL
25mL
7581-97-7
100.0
Glass bottle
Liquid
C4H8Cl2
127.01
01, 119
≥97.5%
Clear colorless to slightly yellow
mixture of DL and meso isomers
97.5
Authentic
1.4410 to 1.4430 (20°C, 589nm)
98%
CH3CH(Cl)CH(Cl)CH3
MFCD00000866
2, 3-Dichlorobutane, 98%

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-486-4

TSCA : TSCA