missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichlorobenzylamine, 97%

Manufacturer:  Alfa Aesar™ L1188403

 View more versions of this product

Catalog No. AAL1188403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2,3-Dichlorobenzylamine
124°C to 126°C (15mmHg)
39226-95-4
MFCD00047927
2639296
JHBVZGONNIVXFJ-UHFFFAOYSA-N
(2,3-dichlorophenyl)methanamine
587625
97%
105°C (221°F)
1g
C7H7Cl2N
UN2735
2,3-dichlorobenzylamine, 2,3-dichlorophenyl methanamine, 2,3-dichloro-benzylamine, 2,3-dichlorobenzyl amine, chembl13165, 1-2,3-dichlorophenyl methanamine, benzenemethanamine, 2,3-dichloro, 2,3-dichlorophenyl methylamine, pubchem16713, acmc-1ae0r
C1=CC(=C(C(=C1)Cl)Cl)CN
176.04
176.05

Chemical Identifiers

39226-95-4
176.04
JHBVZGONNIVXFJ-UHFFFAOYSA-N
587625
C1=CC(=C(C(=C1)Cl)Cl)CN
C7H7Cl2N
MFCD00047927
2,3-dichlorobenzylamine, 2,3-dichlorophenyl methanamine, 2,3-dichloro-benzylamine, 2,3-dichlorobenzyl amine, chembl13165, 1-2,3-dichlorophenyl methanamine, benzenemethanamine, 2,3-dichloro, 2,3-dichlorophenyl methylamine, pubchem16713, acmc-1ae0r
(2,3-dichlorophenyl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Documents