Learn More
2,3-Dichlorobenzonitrile, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1480914
Description
2,3-Dichlorobenzonitrile, is used as a reactant involved in nucleophilic aromatic fluorination, reactions with magnesium amides for synthesis of carboxamides, Suzuki-Miyaura coupling reactions. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3-Dichlorobenzonitrile, is used as a reactant involved in nucleophilic aromatic fluorination, reactions with magnesium amides for synthesis of carboxamides, Suzuki-Miyaura coupling reactions. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.
Solubility
It is miscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
2,3-Dichlorobenzonitrile | |
60°C to 65°C | |
C7H3Cl2N | |
25 g | |
2438753 | |
It is miscible with water. | |
C1=CC(=C(C(=C1)Cl)Cl)C#N | |
172.008 | |
172.01 |
6574-97-6 | |
114°C (6 mmHg) | |
MFCD00016372 | |
UN3439 | |
benzonitrile, dichloro, 2,3-dichlorobenzenecarbonitrile, benzonitrile, 2,3-dichloro, dichlorobenzonitrile, pubchem12516, 2,3-dichloro benzonitrile, 2,3-dichlorophenyl cyanide, 2,3-di-chloro-benzonitrile, acmc-1b38y, ksc357s4b | |
OHDYZVVLNPXKDX-UHFFFAOYSA-N | |
2,3-dichlorobenzonitrile | |
736567 | |
98% |
Chemical Identifiers
6574-97-6 | |
172.008 | |
OHDYZVVLNPXKDX-UHFFFAOYSA-N | |
736567 | |
C1=CC(=C(C(=C1)Cl)Cl)C#N |
C7H3Cl2N | |
MFCD00016372 | |
benzonitrile, dichloro, 2,3-dichlorobenzenecarbonitrile, benzonitrile, 2,3-dichloro, dichlorobenzonitrile, pubchem12516, 2,3-dichloro benzonitrile, 2,3-dichlorophenyl cyanide, 2,3-di-chloro-benzonitrile, acmc-1b38y, ksc357s4b | |
2,3-dichlorobenzonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only