missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichlorobenzenesulfonyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1228306

 View more versions of this product

Catalog No. AAA1228306



Specifications

Specifications

2,3-Dichlorobenzenesulfonyl chloride
57°C to 62°C
82417-45-6
MFCD00051844
7703966
PQODWTNHDKDHIW-UHFFFAOYSA-N
2,3-dichlorobenzenesulfonyl chloride
2734222
98%
131°C to 133°C (1mmHg)
5g
C6H3Cl3O2S
UN3261
2,3-dichlorobenzene-1-sulfonyl chloride, 2,3-dichlorobenzenesulphonyl chloride, 2,3-dichloro-benzenesulfonyl chloride, 2,3-dichlorobenzenesulphonylchloride, 2,3-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 2,3-dichloro-9ci, 2,3-dichlorobenzenesulfonylchloride, pubchem5114, buttpark 9357-48, acmc-1bl9r
C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl
245.498
245.51

Chemical Identifiers

82417-45-6
245.498
PQODWTNHDKDHIW-UHFFFAOYSA-N
2734222
C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl
C6H3Cl3O2S
MFCD00051844
2,3-dichlorobenzene-1-sulfonyl chloride, 2,3-dichlorobenzenesulphonyl chloride, 2,3-dichloro-benzenesulfonyl chloride, 2,3-dichlorobenzenesulphonylchloride, 2,3-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 2,3-dichloro-9ci, 2,3-dichlorobenzenesulfonylchloride, pubchem5114, buttpark 9357-48, acmc-1bl9r
2,3-dichlorobenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

SDS Documents