missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichlorobenzenesulfonyl chloride, 97%, ACROS Organics™

Chemical Identifiers

CAS 82417-45-6
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.51
MDL Number MFCD00051844
InChI Key PQODWTNHDKDHIW-UHFFFAOYSA-N
Synonym 2,3-dichloro-benzenesulfonyl chloride
PubChem CID 2734222
IUPAC Name 2,3-dichlorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC356600050 SDS View Documents Thermo Fisher Scientific
AC356600050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

82417-45-6
245.51
PQODWTNHDKDHIW-UHFFFAOYSA-N
2734222
C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl
C6H3Cl3O2S
MFCD00051844
2,3-dichloro-benzenesulfonyl chloride
2,3-dichlorobenzenesulfonyl chloride

Specifications

Authentic
140.0°C to 142.0°C (2.0mmHg)
61.0°C to 65.0°C
82417-45-6
100.0
C6H3Cl3O2S
MFCD00051844
PQODWTNHDKDHIW-UHFFFAOYSA-N
2,3-dichlorobenzenesulfonyl chloride
2734222
Powder, Crystals or Chunks
2, 3-Dichlorobenzenesulfonyl chloride
Glass bottle
Tan to White
5g
96.0
96% min. (ex Cl) (Argentometry)
Cl(Cl)C6H3SO2Cl
2,3-dichloro-benzenesulfonyl chloride
C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl
245.51
245.51
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger