Learn More
2,3-Dichlorobenzeneboronic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2278114
Description
2,3-Dichlorobenzeneboronic acid is used as a reactant for Suzuki-Miyaura coupling reactions. It is also used in preparation of biologically and pharmacologically active molecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3-Dichlorobenzeneboronic acid is used as a reactant for Suzuki-Miyaura coupling reactions. It is also used in preparation of biologically and pharmacologically active molecules.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
2,3-Dichlorobenzeneboronic acid | |
233°C to 242°C | |
MFCD01075703 | |
2,3-dichlorophenyl boronic acid, 2,3-dichlorobenzeneboronic acid, 2,3-dichlorophenyl boranediol, boronic acid, 2,3-dichlorophenyl, contains varying amounts of anhydride, pubchem1810, dichlorophenylboronic acid, acmc-1c0ur, dichlorobenzene boronic acid | |
TYIKXPOMOYDGCS-UHFFFAOYSA-N | |
(2,3-dichlorophenyl)boronic acid | |
2734661 | |
98% |
151169-74-3 | |
C6H5BCl2O2 | |
25 g | |
Soluble in methanol. | |
OB(O)C1=C(Cl)C(Cl)=CC=C1 | |
190.81 | |
190.82 |
Chemical Identifiers
151169-74-3 | |
190.81 | |
TYIKXPOMOYDGCS-UHFFFAOYSA-N | |
2734661 | |
OB(O)C1=C(Cl)C(Cl)=CC=C1 |
C6H5BCl2O2 | |
MFCD01075703 | |
2,3-dichlorophenyl boronic acid, 2,3-dichlorobenzeneboronic acid, 2,3-dichlorophenyl boranediol, boronic acid, 2,3-dichlorophenyl, contains varying amounts of anhydride, pubchem1810, dichlorophenylboronic acid, acmc-1c0ur, dichlorobenzene boronic acid | |
(2,3-dichlorophenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only