missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichlorobenzaldehyde, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC232675000

 View more versions of this product

Catalog No. AC232675000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 3-Dichlorobenzaldehyde
135°C
Glass bottle
White
500g
98.5
98.5% min. (GC)
Cl2C6H3CHO
07, III, 878
Solubility in water: slightly soluble
C1=CC(=C(C(=C1)Cl)Cl)C=O
175.01
175.01
99%
135g/mL
Authentic
144.0°C (30.0 mmHg)
60.0°C to 64.0°C
6334-18-5
100.0
C7H4Cl2O
MFCD00010127
2,3-dichloro-benzaldehyde, 2,3-dichlorobenzaldehyd, acmc-209nf0, benzaldehyde, 2,3-dichloro, benzaldehyde, dichloro, benzaldehyde,3-dichloro, ccris 8612, dichlorbenzaldehyd, dichlorobenzaldehyde, pubchem3079
LLMLNAVBOAMOEE-UHFFFAOYSA-N
2,3-dichlorobenzaldehyde
35745
Crystalline Powder

Chemical Identifiers

6334-18-5
175.01
LLMLNAVBOAMOEE-UHFFFAOYSA-N
35745
C1=CC(=C(C(=C1)Cl)Cl)C=O
C7H4Cl2O
MFCD00010127
2,3-dichloro-benzaldehyde, 2,3-dichlorobenzaldehyd, acmc-209nf0, benzaldehyde, 2,3-dichloro, benzaldehyde, dichloro, benzaldehyde,3-dichloro, ccris 8612, dichlorbenzaldehyd, dichlorobenzaldehyde, pubchem3079
2,3-dichlorobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 228-711-3

SDS Documents